AC5424073313 Accela ChemBio Inc.

Ethyl 2-Cyanobutanoate

  • MDL number MFCD02258598
  • Cas Number : 1619-58-5
  • Molecular Weight : 141.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005851&attname=GoodsCode

Safety Information

signalword Warning
pcodes P210-P261-P280-P305+P351+P338-P405-P501A
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY005851.pdf
hstatements H302-H312-H332-H315-H319-H335-H227
Availability details for
Quantity
  • Availability
  • Add to Cart