AC5424073419 Accela ChemBio Inc.

2-(4-Methyl-1-piperazinyl)-4-(methylsulfonyl)benzoic Acid

  • MDL number MFCD13191616
  • Cas Number : 1197193-10-4
  • Molecular Weight : 298.35802

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006927&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY006927.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart