AC5424073438 Accela ChemBio Inc.

Methyl N-Boc-2-piperidineacetate

  • MDL number MFCD02179005
  • Cas Number : 183859-36-1
  • Molecular Weight : 257.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006982&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY006982.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart