AC5424073603 Accela ChemBio Inc.

3-Chloro-6-piperazinopyridazine

  • MDL number MFCD05858981
  • Cas Number : 56392-83-7
  • Molecular Weight : 198.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008584&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY008584.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart