AC5424073613 Accela ChemBio Inc.

1-Acetyl-4-methylenepiperidine

  • MDL number MFCD00831797
  • Cas Number : 308087-58-3
  • Molecular Weight : 139.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008656&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY008656.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart