AC5424073617 Accela ChemBio Inc.

2-Chloro-4-methylnicotinamide

  • MDL number MFCD09033826
  • Cas Number : 152362-01-1
  • Molecular Weight : 170.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008695&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY008695.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart