AC5424073673 Accela ChemBio Inc.

5-Bromo-4-isopropylthiazole

  • MDL number MFCD18070981
  • Cas Number : 1025700-46-2
  • Molecular Weight : 206.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY010924&attname=GoodsCode

Safety Information

hcodes No GHS SYMBOl
MSDS http://pdf.accelachem.com/cn/msds/SY010924.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart