AC5424073682 Accela ChemBio Inc.

2-(5-Nitro-2-pyridinyl)acetonitrile

  • MDL number MFCD09033814
  • Cas Number : 123846-66-2
  • Molecular Weight : 163.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011019&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY011019.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart