AC5424073690 Accela ChemBio Inc.

Methyl 2-Methyl-3-(methylsulfonyl)benzoate

  • MDL number MFCD18071000
  • Cas Number : 1256633-15-4
  • Molecular Weight : 228.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011045&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY011045.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart