AC5424073695 Accela ChemBio Inc.

Methyl 2-(Methylsulfonamido)phenylacetate

  • MDL number MFCD18071005
  • Cas Number : 117239-82-4
  • Molecular Weight : 243.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011085&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY011085.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart