AC5424073700 Accela ChemBio Inc.

3,5-Difluoro-4-morpholinophenylboronic Acid Pinacol Ester

  • MDL number MFCD18375252
  • Cas Number : 1313738-69-0
  • Molecular Weight : 325.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011323&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY011323.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart