AC5424073714 Accela ChemBio Inc.

Methyl N-Boc-3-aminophenylacetate

  • MDL number MFCD18071010
  • Cas Number : 1256633-22-3
  • Molecular Weight : 265.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011631&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY011631.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart