AC5424073736 Accela ChemBio Inc.

Ytterbium(III) Sulfate Octahydrate

  • MDL number MFCD00149887
  • Cas Number : 10034-98-7
  • Molecular Weight : 778.39004

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99.9
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011843&attname=GoodsCode

Safety Information

pcodes P261-P280-P305+P351+P338-P304+P340-P405-P501A
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY011843.pdf
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart