AC5424073761 Accela ChemBio Inc.

L-Homophenylalanine tert-Butyl Ester Hydrochloride

  • MDL number MFCD09029247
  • Cas Number : 130316-46-0
  • Molecular Weight : 271.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012064&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012064.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart