AC5424073782 Accela ChemBio Inc.

3-(1H-Tetrazol-5-yl)phenol

  • MDL number MFCD06797247
  • Cas Number : 96859-34-6
  • Molecular Weight : 162.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012314&attname=GoodsCode

Safety Information

pcodes P261-P280-P305+P351+P338-P304+P340-P405-P501A
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY012314.pdf
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart