AC5424073792 Accela ChemBio Inc.

Ethyl 4,5-Difluoro-2-nitrobenzoate

  • MDL number MFCD11042826
  • Cas Number : 1214387-06-0
  • Molecular Weight : 231.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012357&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY012357.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart