AC5424073811 Accela ChemBio Inc.

4-(1-Methyl-5-imidazolyl)phenol

  • MDL number MFCD18375274
  • Cas Number : 74730-75-9
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012405&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY012405.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart