AC5424073827 Accela ChemBio Inc.

5-(4-Bromo-2-fluorophenyl)-2H-tetrazole

  • MDL number MFCD04035676
  • Cas Number : 530081-35-7
  • Molecular Weight : 243.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012445&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY012445.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart