AC5424073829 Accela ChemBio Inc.

5-[4-(Methylsulfonyl)phenyl]-2H-tetrazole

  • MDL number MFCD07432784
  • Cas Number : 441054-54-2
  • Molecular Weight : 224.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012447&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY012447.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart