AC5424073839 Accela ChemBio Inc.

2-(3-Nitrophenyl)imidazole

  • MDL number MFCD05863370
  • Cas Number : 13682-18-3
  • Molecular Weight : 189.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012459&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012459.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart