AC5424073844 Accela ChemBio Inc.

2-(2-Nitrophenyl)imidazole

  • MDL number MFCD05863372
  • Cas Number : 4205-06-5
  • Molecular Weight : 189.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012465&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY012465.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart