AC5424073845 Accela ChemBio Inc.

4-(Imidazol-2-yl)benzonitrile

  • MDL number MFCD05863386
  • Cas Number : 98298-49-8
  • Molecular Weight : 169.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012466&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY012466.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart