AC5424073855 Accela ChemBio Inc.

5-Amino-3-(2-chlorophenyl)isoxazole

  • MDL number MFCD06199356
  • Cas Number : 27025-74-7
  • Molecular Weight : 194.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012493&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY012493.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart