AC5424073864 Accela ChemBio Inc.

5-Amino-3-(3,4-dichlorophenyl)pyrazole Hydrochloride

  • MDL number MFCD06653328
  • Cas Number : 1025447-55-5
  • Molecular Weight : 264.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012515&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY012515.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart