AC5424073874 Accela ChemBio Inc.

2-Chloro-3-nitrobenzamide

  • MDL number MFCD00482025
  • Cas Number : 117054-76-9
  • Molecular Weight : 200.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012811&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY012811.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart