AC5424073875 Accela ChemBio Inc.

2-[Bis(methylsulfonyl)amino]benzonitrile

  • MDL number MFCD01648439
  • Cas Number : 235100-91-1
  • Molecular Weight : 274.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012814&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012814.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart