AC5424073888 Accela ChemBio Inc.

Methyl 4-Chloro-2,6-dinitrobenzoate

  • MDL number MFCD18651711
  • Cas Number : 100418-47-1
  • Molecular Weight : 260.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013036&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY013036.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart