AC5424073911 Accela ChemBio Inc.

(R)-3-Hydroxy-N,N-dimethylpyrrolidine-1-carboxamide

  • MDL number MFCD18651719
  • Cas Number : 1315054-87-5
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Colorless to brown liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013251&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY013251.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart