AC5424073953 Accela ChemBio Inc.

Ethyl 2-(2-Methylimidazol-1-yl)acetate

  • MDL number MFCD03248721
  • Cas Number : 239065-60-2
  • Molecular Weight : 168.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013335&attname=GoodsCode

Safety Information

hcodes No GHS SYMBOl
MSDS http://pdf.accelachem.com/cn/msds/SY013335.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart