AC5424073956 Accela ChemBio Inc.

O-Isobutylhydroxylamine Hydrochloride

  • MDL number MFCD00060208
  • Cas Number : 6084-58-8
  • Molecular Weight : 125.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013358&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY013358.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart