AC5424073981 Accela ChemBio Inc.

5-(3-Phenoxyphenyl)-1H-tetrazole

  • MDL number MFCD02093975
  • Cas Number : 374538-02-0
  • Molecular Weight : 238.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013586&attname=GoodsCode

Safety Information

signalword Warning
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY013586.pdf
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart