AC5424073984 Accela ChemBio Inc.

Methyl 3-Nitropyridine-2-carboxylate

  • MDL number MFCD09953158
  • Cas Number : 103698-08-4
  • Molecular Weight : 182.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013590&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY013590.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart