AC5424073985 Accela ChemBio Inc.

3-Chloro-5-nitropyridine

  • MDL number MFCD04114218
  • Cas Number : 22353-33-9
  • Molecular Weight : 158.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013592&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY013592.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart