AC5424073987 Accela ChemBio Inc.

Ethyl 1-Boc-3-(1-phenylethylamino)piperidine-4-carboxylate

  • MDL number MFCD18911735
  • Cas Number : 1016259-54-3
  • Molecular Weight : 376.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013596&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY013596.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart