AC5424073990 Accela ChemBio Inc.

2-(2-Naphthyloxy)benzonitrile

  • MDL number MFCD12618618
  • Cas Number : 1041593-26-3
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013635&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY013635.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart