AC5424073994 Accela ChemBio Inc.

8-Cbz-2,4-dioxo-1,3,8-triazaspiro[4.5]decane

  • MDL number MFCD18263009
  • Cas Number : 28121-73-5
  • Molecular Weight : 303.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY013644&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY013644.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart