AC5424074042 Accela ChemBio Inc.

3-(3-Indolyl)-2-methylpropionic Acid

  • MDL number MFCD01860840
  • Cas Number : 15142-91-3
  • Molecular Weight : 203.23712

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014277&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY014277.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart