AC5424074073 Accela ChemBio Inc.

2-(Boc-amino)-2-(1,2,3,4-tetrahydro-2-naphthyl)acetic Acid

  • MDL number MFCD04115525
  • Cas Number : 936214-27-6
  • Molecular Weight : 305.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014439&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY014439.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart