AC5424074098 Accela ChemBio Inc.

1-(3-Pyridyl)-1-butylamine

  • MDL number MFCD01691643
  • Cas Number : 90565-27-8
  • Molecular Weight : 150.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014556&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY014556.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart