AC5424074127 Accela ChemBio Inc.

3-(Hydroxymethyl)oxetane-3-carboxylic Acid

  • MDL number MFCD20040523
  • Cas Number : 1379325-60-6
  • Molecular Weight : 132.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014729&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY014729.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart