AC5424074128 Accela ChemBio Inc.

Ethyl 3-Fluoropyridine-2-carboxylate

  • MDL number MFCD11616884
  • Cas Number : 1187732-69-9
  • Molecular Weight : 169.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014737&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY014737.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart