AC5424074143 Accela ChemBio Inc.

Ethyl 1-Benzylimidazole-2-carboxylate

  • MDL number MFCD11857953
  • Cas Number : 865998-45-4
  • Molecular Weight : 230.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014808&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY014808.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart