AC5424074182 Accela ChemBio Inc.

5-Bromo-2-(methylsulfonamido)benzoic Acid

  • MDL number MFCD00452520
  • Cas Number : 1225713-29-0
  • Molecular Weight : 294.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY014997&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY014997.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart