AC5424074192 Accela ChemBio Inc.

3-Fluoro-4-(methylsulfonamido)benzoic Acid

  • MDL number MFCD18285558
  • Cas Number : 716361-59-0
  • Molecular Weight : 233.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015010&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY015010.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart