AC5424074229 Accela ChemBio Inc.

6-(2-Methoxyphenyl)nicotinic Acid

  • MDL number MFCD06410400
  • Cas Number : 887976-03-6
  • Molecular Weight : 229.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015511&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY015511.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart