AC5424074239 Accela ChemBio Inc.

6-(2-Fluorophenyl)-3-pyridinecarbaldehyde

  • MDL number MFCD04115443
  • Cas Number : 898795-78-3
  • Molecular Weight : 201.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015525&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015525.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart