AC5424074244 Accela ChemBio Inc.

2,3-Dihydro-3-methyl-1,2-benzisothiazole 1,1-Dioxide

  • MDL number MFCD19442797
  • Cas Number : 84108-98-5
  • Molecular Weight : 183.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015537&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015537.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart