AC5424074261 Accela ChemBio Inc.

2-Chloro-6-nitroaniline

  • MDL number MFCD00179568
  • Cas Number : 769-11-9
  • Molecular Weight : 172.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015647&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015647.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart