AC5424074297 Accela ChemBio Inc.

2-Bromo-9,9-dibutylfluorene

  • MDL number MFCD12031333
  • Cas Number : 88223-35-2
  • Molecular Weight : 357.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015798&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY015798.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart