AC5424074304 Accela ChemBio Inc.

3-(Fmoc-amino)-3-(1-Boc-3-piperidyl)propanoic Acid

  • MDL number MFCD03425249
  • Cas Number : 372144-11-1
  • Molecular Weight : 494.57936

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015836&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY015836.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart